Please login to your online account to display your discounted pricing

YM 511, Tocris

Potent aromatase (CYP19) inhibitor

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material YM 511
Assay Percent Range >99%
Molecular Formula C16H12BrN5
Formula Weight 354.2
Synonym 4-[[(4-Bromophenyl)methyl]-4H-1, 2, 4-triazol-4-ylamino]benzonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

327850

 
r & d systems
327850
50mg Each for $844.73

327810

 
r & d systems
327810
10mg Each for $201.36
Description & Specifications

Specifications

Formula Weight 354.2
Quantity 50mg
Chemical Name or Material YM 511
Assay Percent Range >99%
Molecular Formula C16H12BrN5
CAS 148869-05-0
Synonym 4-[[(4-Bromophenyl)methyl]-4H-1, 2, 4-triazol-4-ylamino]benzonitrile
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature