Please login to your online account to display your discounted pricing

YM 022, Tocris

Highly potent, selective non-peptide CCK2 antagonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material YM 022
Assay Percent Range >99%
Molecular Formula C32H28N4O3
Formula Weight 516.6
Synonym (R)-N-[2, 3-Dihydro-1-[2-(2-methylphenyl)-2-oxoethyl]-2-oxo-5-phenyl-1H-1, 4-benzodiazepin-3-yl]-N'-(3-methylphenyl)-urea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

140850

 
r & d systems
140850
50mg Each for $976.15

140810

 
r & d systems
140810
10mg Each for $239.63
Description & Specifications

Specifications

Formula Weight 516.6
Quantity 50mg
Chemical Name or Material YM 022
Assay Percent Range >99%
Molecular Formula C32H28N4O3
CAS 145084-28-2
Synonym (R)-N-[2, 3-Dihydro-1-[2-(2-methylphenyl)-2-oxoethyl]-2-oxo-5-phenyl-1H-1, 4-benzodiazepin-3-yl]-N'-(3-methylphenyl)-urea
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature