Please login to your online account to display your discounted pricing

YIL 781, Tocris™

Ghrelin receptor (GHS-R1a) antagonist

$270.88 - $1,140.36

Chemical Identifiers

Chemical Name or Material YIL 781
Assay Percent Range >98%
Molecular Formula C24H28FN3O2·HCl
Formula Weight 445.96
Synonym 6-(4-Fluorophenoxy)-2-methyl-3-[[(3S)-1-(1-methylethyl)-3-piperidinyl]methyl]-4(3H)-quinazolinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

395910

 
r & d systems
RDS395910
10mg Each for $270.88

395950

 
r & d systems
RDS395950
50mg Each for $1,140.36
Description & Specifications

Specifications

Formula Weight 445.96
Quantity 10mg
Chemical Name or Material YIL 781
Assay Percent Range >98%
Molecular Formula C24H28FN3O2·HCl
CAS 875258-85-8
Synonym 6-(4-Fluorophenoxy)-2-methyl-3-[[(3S)-1-(1-methylethyl)-3-piperidinyl]methyl]-4(3H)-quinazolinone
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature