Please login to your online account to display your discounted pricing

Yeast Extract, Powder, MP Biomedicals™


Quantity 250g

Manufacturer: mp biomedicals  0219402783

 View more versions of this product

Catalog No. ICN19402783

  • / Each
 

Description & Specifications

Chemical Identifiers

Appearance Powder
Physical State Solid
Quantity 250g
Chemical Name or Material Yeast Extract
CAS 8013-01-2
Purity Grade Molecular Biology Reagent
Physical Form Yellow-tan powder
MDL Number MFCD00132599
EINECS Number 232-387-9

Specifications

Chemical Name or Material Yeast Extract
CAS 8013-01-2
Purity Grade Molecular Biology Reagent
Physical Form Yellow-tan powder
MDL Number MFCD00132599