Please login to your online account to display your discounted pricing

Y-27632 dihydrochloride, Tocris

Selective p160ROCK inhibitor

$137.56 - $1,333.44

Chemical Identifiers

Chemical Name or Material Y-27632 dihydrochloride
Assay Percent Range >98%
Molecular Formula C14H21N3O·2HCl
Formula Weight 320.26
Synonym trans-4-[(1R)-1-Aminoethyl]-N-4-pyridinylcyclohexanecarboxamide dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

125410

 
r & d systems
RDS125410
10mg Each for $325.38

12541

 
r & d systems
RDS12541
1mg Each for $137.56

125450

 
r & d systems
RDS125450
50mg Each for $1,333.44
Description & Specifications

Specifications

Formula Weight 320.26
Quantity 10mg
Chemical Name or Material Y-27632 dihydrochloride
CAS 129830-38-2
Assay Percent Range >98%
Molecular Formula C14H21N3O·2HCl
Synonym trans-4-[(1R)-1-Aminoethyl]-N-4-pyridinylcyclohexanecarboxamide dihydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature