Please login to your online account to display your discounted pricing

Y-27152, Tocris

Prodrug of Kir6 (KATP) channel opener Y-26763; orally active in vivo

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material Y-27152
Assay Percent Range >98%
Molecular Formula C21H22N2O4
Formula Weight 366.41
Synonym N-[(3S,4R)-6-Cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2H-1-benzopyran-4-yl]-N-(phenylmethoxy)acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

207710

 
r & d systems
RDS207710
10mg Each for $257.85

207750

 
r & d systems
RDS207750
50mg Each for $1,073.10
Description & Specifications

Specifications

Formula Weight 366.41
Quantity 10mg
Chemical Name or Material Y-27152
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C21H22N2O4
CAS 127408-30-4
Synonym N-[(3S,4R)-6-Cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2H-1-benzopyran-4-yl]-N-(phenylmethoxy)acetamide
Safety and Handling

Store at Room Temperature