Please login to your online account to display your discounted pricing

Y 11, Tocris™

Potent and selective FAK inhibitor

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material Y 11
Assay Percent Range >98%
Molecular Formula C8H17N4OBr
Formula Weight 265.15
Synonym 1-(2-Hydroxyethyl)-3,5,7-triaza-1-azaniatricyclo[3.3.1.13,7]decane bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

449850

 
r & d systems
449850
50mg Each for $844.73

449810

 
r & d systems
449810
10mg Each for $201.36
Description & Specifications

Specifications

Formula Weight 265.15
Quantity 50mg
Chemical Name or Material Y 11
Assay Percent Range >98%
CAS 1086639-59-9
Recommended Storage Store at -20°C
Molecular Formula C8H17N4OBr
Synonym 1-(2-Hydroxyethyl)-3,5,7-triaza-1-azaniatricyclo[3.3.1.13,7]decane bromide
Safety and Handling

Store at -20°C