missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Xylitol, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC225985000

 View more versions of this product

Catalog No. AC225985000



Description

Description

Specifications

Specifications

Xylitol
Plastic bottle
0.5% max.
White
500g
87-99-0
C5H12O5
MFCD00064292
15, 10283
Solubility in water: 1700g/L (20°C). Other solubilities: soluble in pyridine,less soluble in alcohols
C(C(C(C(CO)O)O)O)O
152.15
152.15
99+%
Authentic
0.5% max.
215.0°C to 217.0°C
92.0°C to 96.0°C
99+%
2% max. Other polyols (GC)
HOCH2[CH(OH)]3CH2OH
01, 531
adonit, adonite, adonitol, d-ribitol, d-xylitol, eutrit, ribitol, xylit, xylite, xylitol
HEBKCHPVOIAQTA-NGQZWQHPSA-N
(2S,4R)-pentane-1,2,3,4,5-pentol
6912
Crystalline Powder

Chemical Identifiers

87-99-0
152.15
HEBKCHPVOIAQTA-NGQZWQHPSA-N
6912
C(C(C(C(CO)O)O)O)O
C5H12O5
MFCD00064292
adonit, adonite, adonitol, d-ribitol, d-xylitol, eutrit, ribitol, xylit, xylite, xylitol
(2S,4R)-pentane-1,2,3,4,5-pentol
Safety and Handling

Safety and Handling

HYGROSCOPIC

EINECSNumber : 201-788-0

RTECSNumber : ZF0800000

TSCA : TSCA

SDS Documents