Please login to your online account to display your discounted pricing

Xylenol Orange TS, 0.1% (w/v) in Alcohol, Ricca Chemical

$256.18 - $256.18

Chemical Identifiers

Chemical Name or Material Xylenol Orange
Name Note Test Solution
CAS 64-17-5
CAS Min % 98.0
CAS Max % 100.0
CAS 3618-43-7
CAS Min % 0.12
CAS Max % 0.13
CAS 64-17-5 (Ethyl Alcohol (Ethanol))
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

938716

 
ricca chemical company
RI938716
Poly natural 500mL Each for $256.18

93874

  SDS
ricca chemical company
RI93874
Poly natural 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear red
Quantity 500mL
Concentration or Composition Notes 0.1% (w/v)
Solvent or Matrix Alcohol
CAS 64-17-5
CAS Min % 98.0
CAS Max % 100.0
CAS Min % 0.12
CAS Max % 0.13
Chemical Name or Material Xylenol Orange
Name Note Test Solution
Packaging Poly natural
CAS 3618-43-7
CAS 64-17-5 (Ethyl Alcohol (Ethanol))