Please login to your online account to display your discounted pricing

Xylenol Orange Sodium Salt, ACS Grade, LabChem™

$198.00 - $198.00

Chemical Identifiers

Chemical Name or Material Xylenol Orange Sodium Salt
CAS 3618-43-7
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C31H28N2Na4O13S
Formula Weight 760.59
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC269907

 
lab chem
LC269907
Amber Glass 5g Each for $198.00
Description & Specifications

Specifications

Formula Weight 760.59
Physical Form Powder
Chemical Name or Material Xylenol Orange Sodium Salt
Quantity 5g
Recommended Storage Room Temperature
CAS 3618-43-7
Health Hazard 2 GHS H Statement
Product is not hazardous.
Linear Formula C31H28N2Na4O13S
Suitability Passes Test
Purity Grade ACS Grade
Packaging Amber Glass
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Identification Passes Test
Melting Point 210°C
Color Dark red to orange
CAS Max % 100
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.

Room Temperature