Please login to your online account to display your discounted pricing

Alfa Aesar™ Xylenol Blue

$24.55 - $65.09

Chemical Identifiers

Chemical Name or Material Xylenol Blue
CAS 125-31-5
Molecular Formula C23H22O5S
Formula Weight 410.49
MDL Number MFCD00005870
Beilstein 363132
Merck Index 14,10083
Synonym p-Xylenolsulfonephthalein
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1745703

  SDS
alfa aesar™
A1745703
1g Each for $24.55

AAA1745706

  SDS
alfa aesar™
A1745706
5g Each for $65.09
Description & Specifications

Specifications

Formula Weight 410.49
Melting Point ≈212°C (dec.)
Quantity 1g
Molecular Formula C23H22O5S
TSCA TSCA
Beilstein 363132
Synonym p-Xylenolsulfonephthalein
Health Hazard 1 Warning
Chemical Name or Material Xylenol Blue
CAS 125-31-5
Merck Index 14,10083
MDL Number MFCD00005870
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.