missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Xylenes, Semiconductor Grade, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 19402K2

 View more versions of this product

Catalog No. AA19402K2



Description

Description

Xylene is used as a solvent in pharmaceutical, printing, rubber, and leather industries. It is an important component of ink, rubber and adhesives as well as in thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels, and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate. It acts as a cleaning agent for steel, silicon wafers and integrated circuits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Xylenes
1330-20-7
Colorless liquid
137°C to 144°C
29°C (84°F)
Aromatic
1.497
MFCD00077264
1901563
106.17
<°C to 25°C
Colorless
0.86
1 L
465°C (869°F)
Liquid
(CH3)2C6H4
10 hPa (8mm Hg) at 20°C
Miscible with aromatic hydrocarbons. Immiscible with water.
Semiconductor

Chemical Identifiers

1330-20-7
(CH3)2C6H4
MFCD00077264
Safety and Handling

Safety and Handling

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c

H226-H304-H312+H332-H315-H319-H335-H373

DOTInformation : Hazard Class: 3, Packing Group: III

EINECSNumber : 215-535-7

RTECSNumber : ZE2100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only