Please login to your online account to display your discounted pricing

Xylenes (Laboratory), Fisher Chemical


Quantity 4L
Packaging Glass Bottle

Manufacturer: fisher scientific company  X44

 View more versions of this product

Catalog No. X4-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 136°C
Formula Weight 106.17
Purity Grade Laboratory Grade
Melting Point −34°C
Quantity 4L
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Molecular Formula C8H10
Color Clear
Vapor Pressure 8.29mmHg at 25°C
Additional Information Vapor Pressure: 8.29mmHg at 25°C
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Harmful if absorbed through skin or if inhaled. Causes eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
Cleanup Symbol Solvent
ChemAlert Storage Symbol Red
MDL Number MFCD00077264
Synonym Dimethylbenzene
Packaging Glass Bottle
Identification Pass Test
CAS 100-41-4
Health Hazard 2 WARNING!
Chemical Name or Material Xylenes
CAS 1330-20-7
Linear Formula C6H4(CH3)2

Specifications

Chemical Name or Material Xylenes
CAS 1330-20-7
CAS 100-41-4
Purity Grade Laboratory Grade
Molecular Formula C8H10
Linear Formula C6H4(CH3)2
Formula Weight 106.17
MDL Number MFCD00077264
Synonym Dimethylbenzene
Accessories

Accessories for Catalog Number: X44

Item Mfr. No. Item Description Price Per Unit Quantity  
296355104
Spilkleen/Cep  CEP LAB SPILL KIT SOLVENTS
spilkleen/cep
CSKLSK1SOL
Each for $341.99
03-439
Eagle™ Thermoplastics™ Safety Bottle Carriers
fisher scientific
B102
Safety bottle carriers are molded-in handle Each for $217.50
Accessories
 

Emergency Overview
Flammable liquid and vapor. Harmful if absorbed through skin or if inhaled. Causes eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0

WARNING!

 

SDS & Safety Documents