Please login to your online account to display your discounted pricing

Xylenes (Laboratory), Fisher Chemical

$261.69 - $881.92

Chemical Identifiers

Chemical Name or Material Xylenes
CAS 1330-20-7
CAS 100-41-4
Purity Grade Laboratory Grade
Molecular Formula C8H10
Linear Formula C6H4(CH3)2
Formula Weight 106.17
MDL Number MFCD00077264
Synonym Dimethylbenzene
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

X4-20

  SDS
fisher scientific company
X420
Steel Pail 20L Each for $881.92

X4-4

  Accessories SDS
fisher scientific company
X44
Glass Bottle 4L Each for $261.69
Case of 4 Each for $1,046.76
Description & Specifications

Specifications

Purity Grade Laboratory Grade
Boiling Point 136°C
Melting Point −34°C
Formula Weight 106.17
Quantity 20L
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Color Clear
Vapor Pressure 8.29mmHg at 25°C
Molecular Formula C8H10
Synonym Dimethylbenzene
CAS 100-41-4
Identification Pass Test
Cleanup Symbol Solvent
MDL Number MFCD00077264
Health Hazard 2 WARNING!
Chemical Name or Material Xylenes
Additional Information Vapor Pressure: 8.29mmHg at 25°C
Packaging Steel Pail
Linear Formula C6H4(CH3)2
CAS 1330-20-7
ChemAlert Storage Symbol Red
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Harmful if absorbed through skin or if inhaled. Causes eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Safety and Handling
 

Emergency Overview
Flammable liquid and vapor. Harmful if absorbed through skin or if inhaled. Causes eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0

WARNING!