Please login to your online account to display your discounted pricing

Xylenes (Histological), Fisher Chemical


Quantity 200L
Packaging Steel Drum

Manufacturer: fisher scientific company  X3S200

 View more versions of this product

Catalog No. X3S-200

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Histological Grade
Boiling Point 136°C
Melting Point −34°C
Formula Weight 106.17
Quantity 200L
CAS Histological Grade
DOT Information DOT Class 3, : Flammable Liquid
Vapor Pressure 8.29mmHg at 25°C
Physical Form Liquid
Color Clear
Molecular Formula C8H10
Identification Pass Test
Synonym Dimethylbenzene
Additional Information Vapor Pressure: 8.29mmHg at 25°C
Packaging Steel Drum
Cleanup Symbol Solvent
MDL Number MFCD00077264
Health Hazard 2 WARNING!
CAS 100-41-4
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Harmful if absorbed through skin or if inhaled. Causes eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
Chemical Name or Material Xylenes
Linear Formula C6H4(CH3)2
CAS 1330-20-7
ChemAlert Storage Symbol Red

Specifications

Chemical Name or Material Xylenes
CAS 1330-20-7
CAS 100-41-4
Purity Grade Histological Grade
Molecular Formula C8H10
Linear Formula C6H4(CH3)2
Formula Weight 106.17
MDL Number MFCD00077264
Synonym Dimethylbenzene
Accessories

Accessories for Catalog Number: X3S200

Item Mfr. No. Item Description Price Per Unit Quantity  
Accessories
 

Emergency Overview
Flammable liquid and vapor. Harmful if absorbed through skin or if inhaled. Causes eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0

WARNING!

 

SDS & Safety Documents