Please login to your online account to display your discounted pricing

Xylenes, contains ethylbenzene, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 4L
Packaging Narrow Mouth PVC Coated Amber Bottle

Manufacturer: avantor performance  949033

 View more versions of this product

Catalog No. 02004576

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 106.17
Chemical Name or Material Xylenes
Molecular Formula C6H4(CH3)2
Quantity 4L
Name Note Contains ethylbenzene
Flash Point 29.00°C
Packaging Narrow Mouth PVC Coated Amber Bottle
Specific Gravity 1L = 0.86kg
CAS 1330-20-7

Specifications

Chemical Name or Material Xylenes
CAS 1330-20-7
Name Note Contains ethylbenzene
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula C6H4(CH3)2
Formula Weight 106.17