Please login to your online account to display your discounted pricing

Xylenes (Certified ACS), Fisher Chemical


Mixture of ortho, meta, and para isomers and may contain some Ethylbenzene

Quantity 4L
Packaging Aluminum Can

Manufacturer: fisher scientific company  X5S4

 View more versions of this product

Catalog No. X5S-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Boiling Point 136°C
Melting Point −34°C
Formula Weight 106.17
Quantity 4L
Purity Grade Notes Assay: ≥98.5%
DOT Information DOT Class 3, : Flammable Liquid
Vapor Pressure 8.29mmHg at 25°C
Purity Grade Certified ACS Reagent Grade
Physical Form Liquid
Color Clear
Molecular Formula C8H10
Sulfur Compounds 0.003% max.
Identification Pass Test
Iron (Fe) 0.1ppm max.
Additional Information Vapor Pressure: 8.29mmHg at 25°C
Packaging Aluminum Can
Silver (Ag) 0.1ppm max.
Cleanup Symbol Solvent
Assay Percent Range Assay: ≥98.5%
MDL Number MFCD00077264
Copper (Cu) 0.1ppm max.
CAS 100-41-4
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Harmful if absorbed through skin or if inhaled. Causes eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
Nickel (Ni) 0.1ppm max.
Residue after Evaporation 0.002% max.
Substances Darkened by H2SO4 Pass Test
Water 0.05% max.
Heavy Metals (as Pb) 0.1ppm max.
Synonym Xylol, Dimethylbenzene
CAS 1330-20-7
ChemAlert Storage Symbol Red
Health Hazard 2 WARNING!
Chemical Name or Material Xylenes

Specifications

Chemical Name or Material Xylenes
CAS 1330-20-7
CAS 100-41-4
Purity Grade Certified ACS Reagent Grade
Assay ≥98.5%
Assay Percent Range Assay: ≥98.5%
Molecular Formula C8H10
Formula Weight 106.17
MDL Number MFCD00077264
Synonym Xylol, Dimethylbenzene
Accessories

Accessories for Catalog Number: X5S4

Item Mfr. No. Item Description Price Per Unit Quantity  
Accessories
 

Emergency Overview
Flammable liquid and vapor. Harmful if absorbed through skin or if inhaled. Causes eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0

WARNING!

 

SDS & Safety Documents