Please login to your online account to display your discounted pricing

Xylenes (Certified ACS), Fisher Chemical


Mixture of ortho, meta, and para isomers and may contain some Ethylbenzene

Quantity 1 gal.
Packaging Poly Bottle

Manufacturer: fisher scientific company  BPX5P1GAL

 View more versions of this product

Catalog No. X5P-1GAL

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 106.17
Boiling Point 136°C
Melting Point −34°C
Quantity 1 gal.
Purity Grade Notes Assay: ≥98.5%
DOT Information DOT Class 3, : Flammable Liquid
Molecular Formula C8H10
Physical Form Liquid
Purity Grade Certified ACS Reagent Grade
Vapor Pressure 8.29mmHg at 25°C
Color Clear
Identification Pass Test
Nickel (Ni) 0.1ppm max.
CAS 100-41-4
Assay Percent Range Assay: ≥98.5%
Substances Darkened by H2SO4 Pass Test
Additional Information Vapor Pressure: 8.29mmHg at 25°C
Heavy Metals (as Pb) 0.1ppm max.
Cleanup Symbol Solvent
MDL Number MFCD00077264
ChemAlert Storage Symbol Red
Water 0.05% max.
Residue after Evaporation 0.002% max.
Sulfur Compounds 0.003% max.
Copper (Cu) 0.1ppm max.
Iron (Fe) 0.1ppm max.
Packaging Poly Bottle
Synonym Xylol, Dimethylbenzene
Silver (Ag) 0.1ppm max.
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Harmful if absorbed through skin or if inhaled. Causes eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
Health Hazard 2 WARNING!
Chemical Name or Material Xylenes
CAS 1330-20-7

Specifications

Chemical Name or Material Xylenes
CAS 1330-20-7
CAS 100-41-4
Purity Grade Certified ACS Reagent Grade
Assay ≥98.5%
Assay Percent Range Assay: ≥98.5%
Molecular Formula C8H10
Formula Weight 106.17
MDL Number MFCD00077264
Synonym Xylol, Dimethylbenzene
Accessories

Accessories for Catalog Number: X5P1GAL

Item Mfr. No. Item Description Price Per Unit Quantity  
296355104
Spilkleen/Cep  CEP LAB SPILL KIT SOLVENTS
spilkleen/cep
CSKLSK1SOL
Each for $341.99
Accessories
 

Emergency Overview
Flammable liquid and vapor. Harmful if absorbed through skin or if inhaled. Causes eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0

WARNING!