Please login to your online account to display your discounted pricing

Xylenes ACS Reagent Grade, Ricca Chemical

$164.06 - $164.06

Chemical Identifiers

Chemical Name or Material Xylenes
CAS 1330-20-7
CAS Min % 98.0
CAS Max % 100.0
Purity Grade ACS Reagent Grade
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RSOX02004C

 
ricca chemical company
RIRSOX02004C
Glass amber 4L Each for $164.06

RSOX02004X4

 
ricca chemical company
RIRSOX02004X4C
Glass amber 16L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 4L
Purity Grade ACS Reagent Grade
Physical Form Liquid
Color Clear colorless
Chemical Name or Material Xylenes
CAS 1330-20-7
CAS Min % 98.0
CAS Max % 100.0
Packaging Glass amber
Shelf Life 60 months
Safety and Handling

60 months