Please login to your online account to display your discounted pricing

Xylenes ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Xylenes
CAS 1330-20-7
Purity Grade ACS Reagent Grade
Molecular Formula C6H4(CH3)2
Formula Weight 106.17
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK86681

 
avantor performance
866806
Glass 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Reagent Grade
Formula Weight 106.17
Quantity 1L
Packaging Glass
CAS 1330-20-7
Molecular Formula C6H4(CH3)2
Chemical Name or Material Xylenes