Please login to your online account to display your discounted pricing

Xylenes, ACS, 98.5+% (Assay, isomers plus ethylbenzene), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Xylenes
CAS 1330-20-7
Purity Grade ACS Grade
Molecular Formula (CH3)2C6H4
Formula Weight 106.17
MDL Number MFCD00077264
UN Number UN1307
Beilstein 1901563
Merck Index 14,10081
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA16371M1

  SDS
alfa aesar
16371M1
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 106.17
Quantity 4x1L
Chemical Name or Material Xylenes
TSCA TSCA
Odor Aromatic
Refractive Index 1.4970
Flash Point 29°C (84°F)
Boiling Point 137° to 144°C
Physical Form Liquid
Melting Point <−25°C
Assay ≥98.5%
Assay Percent Notes (Assay, isomers plus ethylbenzene)
Molecular Formula (CH3)2C6H4
MDL Number MFCD00077264
CAS 1330-20-7
UN Number UN1307
Beilstein 1901563
Merck Index 14,10081
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H312-H332-H315;
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Density 0.860
Health Hazard 3 GHS P Statement
P210-P262-P302+P352;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not get in eyes, on skin, or on clothing.
IF ON SKIN: Wash with plenty of soap and water.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: XYLENES
RTECS Number ZE2100000
Safety and Handling

Warning

GHS P Statement
P210-P262-P302+P352;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not get in eyes, on skin, or on clothing.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H226-H312-H332-H315;
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.