missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Xylenes, ACS, 98.5%, Alfa Aesar

Manufacturer:  Alfa Aesar 16371K2

 View more versions of this product

Catalog No. AA16371K2



Specifications

Specifications

Xylenes
1330-20-7
(CH3)2C6H4
106.17
ACS
Colorless
For ACS analysis
>98.5%
MFCD00077264
Liquid
0.860
1L

Chemical Identifiers

1330-20-7
(CH3)2C6H4
MFCD00077264
Safety and Handling

Safety and Handling

H226-H312-H332-H315-H303: Flammable liquid and vapour. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. May be harmful if swallowed.

DOTInformation : Hazard Class: 3, Packing Group: III

EINECSNumber : 215-535-7

SDS Documents