Learn More
Xylenecyanol FF, dye content 70%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2153009
Description
Xylene Cyanol FF is a tracking dye commonly used for nucleic acids. The compound has been used in both electrophoresis polyacrylamide and agarose gels for various evaluations including DNA sequencing. The compound has also been used in a mobility shift analysis of ssDNA. In addition, two highly sensitive spectrophotometric assays have been developed for the detection of platinum (IV) (sc-250738, Platinum(IV) chloride) and Chromium (sc-214714) with the use of Xylene Cyanol FF. As an organic dye-inactivated Sendai virus vaccine, Xylene Cyanol FF displayed strong protection against the virus.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsXylene Cyanol FF is a tracking dye commonly used for nucleic acids. The compound has been used in both electrophoresis polyacrylamide and agarose gels for various evaluations including DNA sequencing. The compound has also been used in a mobility shift analysis of ssDNA. In addition, two highly sensitive spectrophotometric assays have been developed for the detection of platinum (IV) (sc-250738, Platinum(IV) chloride) and Chromium (sc-214714) with the use of Xylene Cyanol FF. As an organic dye-inactivated Sendai virus vaccine, Xylene Cyanol FF displayed strong protection against the virus.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents.
Specifications
Xylenecyanol FF | |
2650-17-1 | |
C25H27N2NaO6S2 | |
10 g | |
Soluble in water. | |
[Na+].CCNC1=C(C)C=C(C=C1)C(=C1/C=C\C(=N\CC)\C(C)=C1)\C1=C(C=C(C=C1)S([O-])(=O)=O)S(O)(=O)=O | |
538.61 |
dye content 70% | |
∼295°C (decomposition) | |
MFCD00019481 | |
Acid blue 147; C.I. 42135 | |
VVLFAAMTGMGYBS-KRQUPCAFSA-M | |
sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z,4Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate | |
538.61 |
Chemical Identifiers
2650-17-1 | |
538.61 | |
VVLFAAMTGMGYBS-KRQUPCAFSA-M | |
sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z,4Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate |
C25H27N2NaO6S2 | |
MFCD00019481 | |
Acid blue 147; C.I. 42135 | |
[Na+].CCNC1=C(C)C=C(C=C1)C(=C1/C=C\C(=N\CC)\C(C)=C1)\C1=C(C=C(C=C1)S([O-])(=O)=O)S(O)(=O)=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 220-167-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only