missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Xylazine

$172.72 - $732.51

Chemical Identifiers

CAS 7361-61-7
Molecular Formula C12H16N2S
Molecular Weight (g/mol) 220.334
MDL Number MFCD00057908
InChI Key BPICBUSOMSTKRF-UHFFFAOYSA-N
Synonym xylazine, rompun, n-2,6-dimethylphenyl-5,6-dihydro-4h-1,3-thiazin-2-amine, xylazin, narcoxyl, 4h-1,3-thiazin-2-amine, n-2,6-dimethylphenyl-5,6-dihydro, xylzin, xylazinum inn-latin, bayer 1470, xilazina inn-spanish
PubChem CID 5707
IUPAC Name N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine
SMILES CC1=C(C(=CC=C1)C)NC2=NCCCS2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6143006 View Documents Alfa Aesar™
J6143006
5g Each for $172.72
 
AAJ6143014 View Documents Alfa Aesar™
J6143014
25g Each for $732.51
 
Description

Description

Applications
Xylazine is an α2 class of adrenergic receptor agonist. Xylazine is a clonidine analoque that acts on presynaptic and postsynaptic receptors as an a 2-adrenergic agonist.

Solubility
Soluble in methanol at 50mg/ml. Also soluble in dilute acid solutions.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

7361-61-7
220.334
BPICBUSOMSTKRF-UHFFFAOYSA-N
5707
CC1=C(C(=CC=C1)C)NC2=NCCCS2
C12H16N2S
MFCD00057908
xylazine, rompun, n-2,6-dimethylphenyl-5,6-dihydro-4h-1,3-thiazin-2-amine, xylazin, narcoxyl, 4h-1,3-thiazin-2-amine, n-2,6-dimethylphenyl-5,6-dihydro, xylzin, xylazinum inn-latin, bayer 1470, xilazina inn-spanish
N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine

Specifications

White
7361-61-7
MFCD00057908
14,10080
Soluble in methanol at 50mg/ml; Also soluble in dilute acid solutions
CC1=C(C(=CC=C1)C)NC2=NCCCS2
220.334
220.33
Xylazine
5g
C12H16N2S
UN2811
xylazine, rompun, n-2,6-dimethylphenyl-5,6-dihydro-4h-1,3-thiazin-2-amine, xylazin, narcoxyl, 4h-1,3-thiazin-2-amine, n-2,6-dimethylphenyl-5,6-dihydro, xylzin, xylazinum inn-latin, bayer 1470, xilazina inn-spanish
BPICBUSOMSTKRF-UHFFFAOYSA-N
N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine
5707
Powder
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : XJ0776300

Recommended Storage : −30° to −10°C