missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Xylazine hydrochloride

Agonist at the alpha-2 adrenergic receptor

$203.55 - $774.15

Chemical Identifiers

CAS 23076-35-9
Molecular Formula C12H17ClN2S
Molecular Weight (g/mol) 256.792
MDL Number MFCD00058196
InChI Key QYEFBJRXKKSABU-UHFFFAOYSA-N
Synonym xylazine hydrochloride, xylazine hcl, n-2,6-dimethylphenyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, celactal, xylazine chloride, xylazine hydrochloride, bay 1470 hydrochloride, unii-ngc3s0882s, ccris 8685, bay va 1470
PubChem CID 68554
IUPAC Name N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine;hydrochloride
SMILES CC1=C(C(=CC=C1)C)NC2=NCCCS2.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6239406 View Documents Alfa Aesar™
J6239406
5g Each for $203.55
 
AAJ6239414 View Documents Alfa Aesar™
J6239414
25g Each for $774.15
 
Specifications

Chemical Identifiers

23076-35-9
256.792
QYEFBJRXKKSABU-UHFFFAOYSA-N
68554
CC1=C(C(=CC=C1)C)NC2=NCCCS2.Cl
C12H17ClN2S
MFCD00058196
xylazine hydrochloride, xylazine hcl, n-2,6-dimethylphenyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, celactal, xylazine chloride, xylazine hydrochloride, bay 1470 hydrochloride, unii-ngc3s0882s, ccris 8685, bay va 1470
N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine;hydrochloride

Specifications

White
23076-35-9
MFCD00058196
6448471
xylazine hydrochloride, xylazine hcl, n-2,6-dimethylphenyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, celactal, xylazine chloride, xylazine hydrochloride, bay 1470 hydrochloride, unii-ngc3s0882s, ccris 8685, bay va 1470
QYEFBJRXKKSABU-UHFFFAOYSA-N
N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine;hydrochloride
68554
Powder
5g
C12H17ClN2S
UN2811
14,10080
Soluble in water
CC1=C(C(=CC=C1)C)NC2=NCCCS2.Cl
256.792
256.79
Xylazine hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H350-H315-H319-H335
Toxic if swallowed.
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : XJ0776350

Recommended Storage : −30° to −10°C