missing translation for 'onlineSavingsMsg'
Learn More

Xylazine Hydrochloride 98.0+%, TCI America™

Catalog No. X00595G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00595G 5 g
X005925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X00595G Supplier TCI America Supplier No. X00595G
Only null left

Chemical Identifiers

CAS 23076-35-9
Molecular Formula C12H17ClN2S
Molecular Weight (g/mol) 256.79
MDL Number MFCD00058196
InChI Key QYEFBJRXKKSABU-UHFFFAOYSA-N
Synonym xylazine hydrochloride, xylazine hcl, n-2,6-dimethylphenyl-5,6-dihydro-4h-1,3-thiazin-2-amine hydrochloride, celactal, xylazine chloride, bay 1470 hydrochloride, unii-ngc3s0882s, ccris 8685, bay va 1470
PubChem CID 68554
IUPAC Name hydrogen N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine chloride
SMILES [H+].[Cl-].CC1=CC=CC(C)=C1NC1=NCCCS1

Specifications

Color White
Quantity 5 g
Physical Form Crystal-Powder at 20°C
UN Number 2811
Formula Weight 256.79
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Xylazine Hydrochloride
RTECSNumber XJ0776350
TSCA No