Please login to your online account to display your discounted pricing

XRP44X, Tocris

Ras-Net pathway inhibitor

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material XRP44X
Assay Percent Range >99%
Molecular Formula C21H21ClN4O
Formula Weight 380.87
Synonym 1-(3-Chlorophenyl)-4-[(3-methyl-1-phenyl-1H-pyrazol-5-yl)carbonyl]piperazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

310110

 
r & d systems
310110
10mg Each for $244.81

310150

 
r & d systems
310150
50mg Each for $997.23
Description & Specifications

Specifications

Formula Weight 380.87
Quantity 10mg
Chemical Name or Material XRP44X
Assay Percent Range >99%
Molecular Formula C21H21ClN4O
Synonym 1-(3-Chlorophenyl)-4-[(3-methyl-1-phenyl-1H-pyrazol-5-yl)carbonyl]piperazine
Recommended Storage Store at Room Temperature
CAS 729605-21-4
Safety and Handling

Store at Room Temperature