Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ XRCC1 (X-Ray Repair Cross Complementing Protein 1) Ab-3, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ XRCC1 (X-Ray Repair Cross Complementing Protein 1) Ab-3, Mouse Monoclonal Antibody. 

Manufacturer: thermo scientific™ lab vision™  MS1393PABX

 View more versions of this product

Catalog No. MS1393PABX

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2a kappa
Cross Reactivity Human
Immunogen Recombinant human XRCC1 protein
Regulatory Status RUO
Antigen XRCC1 (X-Ray Repair Cross Complementing Protein1) Ab-3
Host Species Mouse
Quantity 200µL
Clone 144
Formulation Ab without BSA/Azide (Frozen)
Purification Method Ls174T, or HeLa cells, normal testis
Research Discipline DNA Repair
Concentration 1mg/mL
Conjugate Unlabeled
Type Primary
Classification Monoclonal
Item Description 200uL; 1mg/mL; Purified w/o BSA, azide

Provide accurate, reproducible results with the Thermo Scientific™ XRCC1 (X-Ray Repair Cross Complementing Protein 1) Ab-3, Mouse Monoclonal Antibody. 

X-Ray Repair Cross Complementing (XRCC1) plays a role in excision repair of DNA after ionizing irradiation. XRCC1 binds to DNA ligase III through the C-terminal 96 amino acids and to DNA polymerase β through the N-terminal half. In testis XRCC1 is expressed at high levels. Cells with a mutation of this gene exhibit decreased single strand break repair and reduced recombination repair. They show increased double strand breaks, and sister chromatid exchange is increased up to 10-fold. The XRCC1 might serve as a scaffold protein during base excision-repair.

Host Species: Mouse

Clone: 144

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human XRCC1 protein

Molecular Weight: 85kDa

Positive Control: Ls174T, or HeLa cells; normal testis

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO