Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ XRCC1 (X-Ray Repair Cross Complementing Protein 1) Ab-3, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ XRCC1 (X-Ray Repair Cross Complementing Protein 1) Ab-3, Mouse Monoclonal Antibody. 

Manufacturer: thermo scientific™ lab vision™  MS1393P1ABX

 View more versions of this product

Catalog No. MS1393P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2a kappa
Regulatory Status RUO
Quantity 100µL
Clone 144
Format Concentrated
Cross Reactivity Human
Host Species Mouse
Immunogen Recombinant human XRCC1 protein
Antigen XRCC1 (X-Ray Repair Cross Complementing Protein1) Ab-3
Conjugate Unlabeled
Research Discipline DNA Repair
Purification Method Ls174T, or HeLa cells, normal testis
Type Primary
Concentration 1mg/mL
Classification Monoclonal
Formulation Ab without BSA/Azide (Frozen)
Item Description 100uL; 1mg/mL; Purified w/o BSA, azide

Provide accurate, reproducible results with the Thermo Scientific™ XRCC1 (X-Ray Repair Cross Complementing Protein 1) Ab-3, Mouse Monoclonal Antibody. 

X-Ray Repair Cross Complementing (XRCC1) plays a role in excision repair of DNA after ionizing irradiation. XRCC1 binds to DNA ligase III through the C-terminal 96 amino acids and to DNA polymerase β through the N-terminal half. In testis XRCC1 is expressed at high levels. Cells with a mutation of this gene exhibit decreased single strand break repair and reduced recombination repair. They show increased double strand breaks, and sister chromatid exchange is increased up to 10-fold. The XRCC1 might serve as a scaffold protein during base excision-repair.

Host Species: Mouse

Clone: 144

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human XRCC1 protein

Molecular Weight: 85kDa

Positive Control: Ls174T, or HeLa cells; normal testis

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO