Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ XPF Ab-5, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ XPF Ab-5, Mouse Monoclonal Antibody.

$268.31 - $1,380.31

Specifications

Antigen XPF Ab-5
Clone 51
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1385P

 
thermo scientific™ lab vision™
MS1385P
200μg/mL 1mL Each for $1,270.22

MS1385P0

 
thermo scientific™ lab vision™
MS1385P0
200μg/mL 100µL Each for $268.31

MS1385P1

 
thermo scientific™ lab vision™
MS1385P1
200μg/mL 500µL Each for $758.94

MS1385P1ABX

 
thermo scientific™ lab vision™
MS1385P1ABX
1mg/mL 100µL Each for $830.47

MS1385PABX

 
thermo scientific™ lab vision™
MS1385PABX
1mg/mL 200µL Each for $1,380.31
Description & Specifications

Specifications

Antigen XPF Ab-5
Clone 51
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline DNA Repair
Type Primary
Immunogen Recombinant human XPF protein
Isotype IgG2b kappa

Provide accurate, reproducible results with the Thermo Scientific™ XPF Ab-5, Mouse Monoclonal Antibody.

The structure-specific ERCC1/XPF endonuclease complex is implicated in the repair of two distinct types of lesions in DNA: NER for UV-induced lesions and bulky chemical adducts; and recombination repair of the very genotoxic interstrand cross-links. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells XPG cleaves 3' of the DNA lesion while the ERCC1-XPF complex makes the 5' incision.

Host Species: Mouse

Clone: 51

Isotype: IgG2b/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human XPF protein

Molecular Weight: ∽110kDa

Positive Control: MCF-7 cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO