Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ XPF Ab-5, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ XPF Ab-5, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1385PABX

 View more versions of this product

Catalog No. MS1385PABX

  • / Each
 

Description & Specifications

Specifications

Applications Western Blotting
Regulatory Status RUO
Cross Reactivity Human
Quantity 200µL
Clone 51
Isotype IgG2b kappa
Format Concentrated
Antigen XPF Ab-5
Host Species Mouse
Immunogen Recombinant human XPF protein
Classification Monoclonal
Type Primary
Research Discipline DNA Repair
Concentration 1mg/mL
Conjugate Unlabeled

Provide accurate, reproducible results with the Thermo Scientific™ XPF Ab-5, Mouse Monoclonal Antibody.

The structure-specific ERCC1/XPF endonuclease complex is implicated in the repair of two distinct types of lesions in DNA: NER for UV-induced lesions and bulky chemical adducts; and recombination repair of the very genotoxic interstrand cross-links. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells XPG cleaves 3' of the DNA lesion while the ERCC1-XPF complex makes the 5' incision.

Host Species: Mouse

Clone: 51

Isotype: IgG2b/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human XPF protein

Molecular Weight: ∽110kDa

Positive Control: MCF-7 cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO