Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ XPF Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ XPF Ab-1, Mouse Monoclonal Antibody.

$300.39 - $1,382.86

Specifications

Antigen XPF Ab-1
Clone 219
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS1381P

 
thermo scientific™ lab vision™
MS1381P
Purified Ab with BSA and Azide 1mL Each for $1,270.88

MS1381P0

 
thermo scientific™ lab vision™
MS1381P0
Purified Ab with BSA and Azide 100µL Each for $300.39

MS1381P1

 
thermo scientific™ lab vision™
MS1381P1
Purified Ab with BSA and Azide 500µL Each for $760.35

MS1381P1ABX

 
thermo scientific™ lab vision™
MS1381P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $832.02

MS1381PABX

 
thermo scientific™ lab vision™
MS1381PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,382.86

MS1381R7

 
thermo scientific™ lab vision™
MS1381R7
Ready-to-use for IHC (UVLP) 7mL Each for $753.80
Description & Specifications

Specifications

Antigen XPF Ab-1
Clone 219
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline DNA Repair
Type Primary
Immunogen Recombinant human XPF protein
Isotype IgG2

Provide accurate, reproducible results with the Thermo Scientific™ XPF Ab-1, Mouse Monoclonal Antibody.

The structure-specific ERCC1/XPF endonuclease complex is implicated in the repair of two distinct types of lesions in DNA: NER for UV-induced lesions and bulky chemical adducts; and recombination repair of the very genotoxic interstrand cross-links. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells XPG cleaves 3' of the DNA lesion while the ERCC1-XPF complex makes the 5' incision.

Host Species: Mouse

Clone: 219

Isotype: IgG2/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human XPF protein

Molecular Weight: ∽110kDa

Positive Control: MCF-7 cells or human tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO