Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ XPA Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ XPA Ab-1, Mouse Monoclonal Antibody.

$268.82 - $1,382.86

Specifications

Antigen XPA Ab-1
Clone 12F5
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS650P

 
thermo scientific™ lab vision™
MS650P
Purified Ab with BSA and Azide 1mL Each for $1,272.57

MS650P0

 
thermo scientific™ lab vision™
MS650P0
Purified Ab with BSA and Azide 100µL Each for $268.82

MS650P1

 
thermo scientific™ lab vision™
MS650P1
Purified Ab with BSA and Azide 500µL Each for $761.65

MS650P1ABX

 
thermo scientific™ lab vision™
MS650P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $832.02

MS650PABX

 
thermo scientific™ lab vision™
MS650PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,382.86

MS650R7

 
thermo scientific™ lab vision™
MS650R7
Ready-to-use for IHC (UVLP) 7mL Each for $817.37
Description & Specifications

Specifications

Antigen XPA Ab-1
Clone 12F5
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline DNA Repair
Type Primary
Immunogen Recombinant human XPA protein
Isotype IgG2a kappa

Provide accurate, reproducible results with the Thermo Scientific™ XPA Ab-1, Mouse Monoclonal Antibody.

The XPA (xeroderma pigmentosum group A) protein specifically recognizes the UV-or chemically damaged DNA lesions, and triggers the nucleotide excision repair process. XPA binds to the replication protein A (RPA) or the excision repair cross complementing 1 protein (ERCC1). In the absence of nucleotide excision repair persisting (unrepaired) DNA lesions (adducts) may lead to the accumulation of gene mutations and ultimately to cancer. Xeroderma pigmentosum patients have a > 2000 fold increased risk to develop skin cancer at sun-exposed areas.

Host Species: Mouse

Clone: 12F5

Isotype: IgG2a/k

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human XPA protein

Molecular Weight: ∽40kDa

Positive Control: Ls174T or MCF-7 cells or human tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO