Please login to your online account to display your discounted pricing

XMD 8-92, Tocris™

Selective ERK5/BMK1 inhibitor

$290.95 - $1,205.80

Chemical Identifiers

Chemical Name or Material XMD 8-92
Assay Percent Range >98%
Molecular Formula C26H30N6O3
Formula Weight 474.55
Synonym 2-[[2-Ethoxy-4-(4-hydroxy-1-piperidinyl)phenyl]amino]-5,11-dihydro-5,11-dimethyl-6H-pyrimido[4,5-b][1,4]benzodiazepin-6-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

413210

 
r & d systems
RDS413210
10mg Each for $290.95

413250

 
r & d systems
RDS413250
50mg Each for $1,205.80
Description & Specifications

Specifications

Formula Weight 474.55
Quantity 10mg
Chemical Name or Material XMD 8-92
Molecular Formula C26H30N6O3
CAS 1234480-50-2
Synonym 2-[[2-Ethoxy-4-(4-hydroxy-1-piperidinyl)phenyl]amino]-5,11-dihydro-5,11-dimethyl-6H-pyrimido[4,5-b][1,4]benzodiazepin-6-one
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Safety and Handling

Store at Room Temperature