Please login to your online account to display your discounted pricing

Xenon difluoride, 99.5% (metals basis), Alfa Aesar™

Fluorinating agent

Chemical Identifiers

Chemical Name or Material Xenon difluoride
CAS 13709-36-9
Molecular Formula XeF2
Formula Weight 169.3
MDL Number MFCD00040538
UN Number UN3087
Merck Index 14,10072
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA3973904

 
alfa aesar
3973904
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 169.3
Melting Point 127° to 129°C
Quantity 2g
Assay 99.5%
Chemical Name or Material Xenon difluoride
TSCA TSCA
Physical Form Crystalline Granules, shipped in teflon
Sensitivity Moisture Sensitive
Assay Percent Notes (metals basis)
Molecular Formula XeF2
MDL Number MFCD00040538
CAS 13709-36-9
UN Number UN3087
Merck Index 14,10072
Color White
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H272-H301-H330-H314-H318;
May intensify fire; oxidizer.
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P221-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a;
Take any precaution to avoid mixing with combustibles.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 4.32
DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
RTECS Number ZE1294116
Safety and Handling

Danger

GHS P Statement
P221-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a;
Take any precaution to avoid mixing with combustibles.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H301-H330-H314-H318;
May intensify fire; oxidizer.
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.