Please login to your online account to display your discounted pricing

XCT 790, Tocris™

Selective ERRα antagonist/inverse agonist

$214.12 - $214.12

Chemical Identifiers

Chemical Name or Material XCT 790
Assay Percent Range >99%
Molecular Formula C23H13F9N4O3S
Formula Weight 596.42
Synonym 3-[4-(2,4-Bis-trifluoromethylbenzyloxy)-3-methoxyphenyl]-2-cyano-N-(5-trifluoromethyl-1,3,4-thiadiazol-2-yl)acrylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

39-281-0

 
r & d systems
RDS392810
10mg Each for $214.12
Description & Specifications

Specifications

Formula Weight 596.42
Quantity 10mg
Chemical Name or Material XCT 790
CAS 725247-18-7
Synonym 3-[4-(2,4-Bis-trifluoromethylbenzyloxy)-3-methoxyphenyl]-2-cyano-N-(5-trifluoromethyl-1,3,4-thiadiazol-2-yl)acrylamide
Recommended Storage Store at +4°C
Assay Percent Range >99%
Molecular Formula C23H13F9N4O3S
Safety and Handling

Store at +4°C