missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Xanthine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1107714
Description
Xanthine and xanthine oxidase system can be used to produce superoxide radicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsXanthine and xanthine oxidase system can be used to produce superoxide radicals.
Solubility
Soluble in water(0.067g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Xanthine | |
>300°C | |
C5H4N4O2 | |
25 g | |
14,10059 | |
Soluble in water(0.067g/L). | |
C1=NC2=C(N1)C(=O)NC(=O)N2 | |
152.113 | |
CHEBI:17712 | |
99% |
69-89-6 | |
Odorless | |
MFCD00078453 | |
8733 | |
xanthine, 2,6-dihydroxypurine, xanthin, 2,6-dioxopurine, pseudoxanthine, 1h-purine-2,6 3h,7h-dione, isoxanthine, xanthic oxide, 1h-purine-2,6-diol, 9h-purine-2,6-diol | |
LRFVTYWOQMYALW-UHFFFAOYSA-N | |
3,7-dihydropurine-2,6-dione | |
1188 | |
152.11 |
Chemical Identifiers
69-89-6 | |
152.113 | |
LRFVTYWOQMYALW-UHFFFAOYSA-N | |
1188 | |
3,7-dihydropurine-2,6-dione |
C5H4N4O2 | |
MFCD00078453 | |
xanthine, 2,6-dihydroxypurine, xanthin, 2,6-dioxopurine, pseudoxanthine, 1h-purine-2,6 3h,7h-dione, isoxanthine, xanthic oxide, 1h-purine-2,6-diol, 9h-purine-2,6-diol | |
CHEBI:17712 | |
C1=NC2=C(N1)C(=O)NC(=O)N2 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 200-718-6
RTECSNumber : ZD7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only