Please login to your online account to display your discounted pricing

Xanthine, 99 to 100%, MP Biomedicals™


Quantity 25g

Manufacturer: mp biomedicals  10329025

 View more versions of this product

Catalog No. ICN-10329025

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Color Tan
Formula Weight 152.1
Quantity 25g
Chemical Name or Material Xanthine
Assay Percent Range 99 to 100%
Molecular Formula C5H4N4O2
CAS 69-89-6
Synonym 2, 6-Dihydroxypurine

Specifications

Chemical Name or Material Xanthine
CAS 69-89-6
Physical Form Powder
Assay Percent Range 99 to 100%
Molecular Formula C5H4N4O2
Formula Weight 152.1
Synonym 2, 6-Dihydroxypurine