Please login to your online account to display your discounted pricing

Xanthine, 99 to 100%, MP Biomedicals™


Quantity 10g

Manufacturer: mp biomedicals  10329010

 View more versions of this product

Catalog No. ICN-10329010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 152.1
Color Tan
Quantity 10g
Synonym 2, 6-Dihydroxypurine
Chemical Name or Material Xanthine
Molecular Formula C5H4N4O2
CAS 69-89-6
Assay Percent Range 99 to 100%

Specifications

Chemical Name or Material Xanthine
CAS 69-89-6
Physical Form Powder
Assay Percent Range 99 to 100%
Molecular Formula C5H4N4O2
Formula Weight 152.1
Synonym 2, 6-Dihydroxypurine