Please login to your online account to display your discounted pricing

Xanthine, 99 to 100%, MP Biomedicals™


Quantity 5g

Manufacturer: mp biomedicals  0210329005

 View more versions of this product

Catalog No. ICN-10329005

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 5g
Chemical Name or Material Xanthine
CAS 69-89-6
Physical Form Tan powder
Assay Percent Range 99 to 100%
Molecular Formula C5H4N4O2
Formula Weight 152.1g/mol
Synonym 2, 6-Dihydroxypurine

Specifications

Chemical Name or Material Xanthine
CAS 69-89-6
Physical Form Tan powder
Assay Percent Range 99 to 100%
Molecular Formula C5H4N4O2
Formula Weight 152.1g/mol
Synonym 2, 6-Dihydroxypurine