Please login to your online account to display your discounted pricing

Xanthine, 99 to 100%, MP Biomedicals™

$66.37 - $145.73

Chemical Identifiers

Chemical Name or Material Xanthine
CAS 69-89-6
Physical Form Powder
Assay Percent Range 99 to 100%
Molecular Formula C5H4N4O2
Formula Weight 152.1
Synonym 2, 6-Dihydroxypurine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN-10329025

 
mp biomedicals
10329025
25g Each for $145.73

ICN-10329010

 
mp biomedicals
10329010
10g Each for $66.37
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 152.1
Color Tan
Quantity 25g
Synonym 2, 6-Dihydroxypurine
Chemical Name or Material Xanthine
Molecular Formula C5H4N4O2
CAS 69-89-6
Assay Percent Range 99 to 100%