Please login to your online account to display your discounted pricing

Xanthene 98% Acros Organics

Chemical Identifiers

Chemical Name or Material Xanthene
CAS 92-83-1
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H10O
Formula Weight 182.22
MDL Number MFCD00005055
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC140951000

  SDS
thermo fisher scientific
AC140951000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 182.22
Assay ≥97.5%
Quantity 100g
Color White to pale yellow
Beilstein 17, 73
Melting Point 100° to 103°C
Chemical Name or Material Xanthene
CAS 92-83-1
Molecular Formula C13H10O
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00005055
Infrared Spectrum Authentic
Assay Percent Range 98%
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.