Please login to your online account to display your discounted pricing

Xanthene 98% Acros Organics


Quantity 100g

Manufacturer: thermo fisher scientific  AC140951000

 View more versions of this product

Catalog No. AC140951000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 182.22
Quantity 100g
Color White to pale yellow
Beilstein 17, 73
Melting Point 100° to 103°C
Chemical Name or Material Xanthene
Molecular Formula C13H10O
MDL Number MFCD00005055
CAS 92-83-1
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Xanthene
CAS 92-83-1
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H10O
Formula Weight 182.22
MDL Number MFCD00005055
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

SDS & Safety Documents