Please login to your online account to display your discounted pricing

Alfa Aesar™ Xanthene-9-carboxamide, 98%

$41.30 - $557.50

Chemical Identifiers

Chemical Name or Material Xanthene-9-carboxamide
CAS 5813-90-1
Molecular Formula C14H11NO2
Formula Weight 225.25
MDL Number MFCD00173894
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1707703

  SDS
alfa aesar™
A1707703
1g Each for $41.30

AAA1707706

  SDS
alfa aesar™
A1707706
5g Each for $144.80

AAA1707714

  SDS
alfa aesar™
A1707714
25g Each for $557.50
Description & Specifications

Specifications

Formula Weight 225.25
Chemical Name or Material Xanthene-9-carboxamide
Assay 98%
Melting Point 244° to 246°C
Molecular Formula C14H11NO2
Quantity 1g
MDL Number MFCD00173894
CAS 5813-90-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.