missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ Xanomeline oxalate

Functionally selective M1 agonist

$218.74 - $915.10

Specifications

Product Type Xanomeline oxalate
Inhibitors Xanomeline oxalate
Molecular Weight 371.46
Purity >98
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
356910R Tocris™
RDS356910
10mg Each for $218.74
356950R Tocris™
RDS356950
50mg Each for $915.10
Specifications

Description

Functionally selective muscarinic M1 receptor agonist (EC50 values are 0.3, 5, 42, 52 and 92.5nM at M1, M3, M5, M4 and M2 receptors respectively). Displays a complex pharmacological profile: reversible and wash-resistant binding, resulting in full agonist activity at M1; delayed wash-resistant partial agonist activity at M2; and delayed wash-resistant full agonist activity at M4. Exhibits antipsychotic activity, and improves cognitive deficits and behavioral disturbances in Alzheimer's disease and schizophrenia.
  • Chemical Name: 3-[4-(Hexyloxy)-1,2,5-thiadiazol-3-yl]-1,2,5,6-tetrahydro-1-methylpyridine oxalate
  • Formula: C19H20BrN3O3
  • CAS Number: 84379-13-5
  • Solubility: Soluble to 100mM in DMSO and to 100mM in ethanol
  • Purity: >98
Description & Specifications

Specifications

Xanomeline oxalate
Xanomeline oxalate
371.46
>98