Please login to your online account to display your discounted pricing

Xaliproden hydrochloride, Tocris

Orally active, high affinity 5-HT1A agonist

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material Xaliproden hydrochloride
Assay Percent Range >99%
Molecular Formula C24H22F3N·HCl
Formula Weight 417.9
Synonym 1,2,3,6-Tetrahydro-1-[2-(2-naphthalenyl)ethyl]-4-[3-(trifluoromethyl)phenyl]-pyridine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

249150

 
r & d systems
RDS249150
50mg Each for $794.96

249110

 
r & d systems
RDS249110
10mg Each for $188.60
Description & Specifications

Specifications

Formula Weight 417.9
Quantity 50mg
Molecular Formula C24H22F3N·HCl
Assay Percent Range >99%
Chemical Name or Material Xaliproden hydrochloride
CAS 90494-79-4
Synonym 1,2,3,6-Tetrahydro-1-[2-(2-naphthalenyl)ethyl]-4-[3-(trifluoromethyl)phenyl]-pyridine hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature