Invitrogen™ Novex™ Value Tris-Glycine Gels DFS Item
Great value-based tris-glycine precast protein gel balancing performance and price.
$211.00 - $211.00
Specifications
Length (Metric) Gel 8cm
Width (Metric) Gel 8cm
Material Tris-Glycine Gel
Thickness (Metric) 1.0mm
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Quantity Price Quantity  
Catalog Number Mfr. No. Description Quantity Price Quantity  
XV00100PK20 Invitrogen™
LSXV00100PK20
10% Value Tris-Glycine Gels, 1.0 mm, 10-well, Box of 20 20 Gels Each for $211.00
XV00105PK20 Invitrogen™
LSXV00105PK20
10% Value Tris-Glycine Gels, 1.0 mm, 15-well, Box of 20 20 Gels Each for $211.00
XV04120PK20 Invitrogen™
LSXV04120PK20
4-12% Value Tris-Glycine Gels, 1.0 mm, 10-well, Box of 20 20 Gels Each for $211.00
XV04125PK20 Invitrogen™
LSXV04125PK20
4-12% Value Tris-Glycine Gels, 1.0 mm, 15-well, Box of 20 20 Gels Each for $211.00
XV04200PK20 Invitrogen™
LSXV04200PK20
Novex™ Value™ 4 to 20% Tris-Glycine Mini Gels, 20 Gels, 0.039 in. Thickness, 8 x 8cm Gel Size, Gel, For Protein Gel Electrophoresis, 12 Months Shelf Life, 2 to 8°C Storage Temperature 20 Gels Each for $211.00
XV04205PK20 Invitrogen™
LSXV04205PK20
4-20% Value Tris-Glycine Gels, 1.0 mm, 10-well, Box of 20 20 Gels Each for $211.00
Specifications
Description
Novex™ Value Tris-Glycine Mini gels provide a great balance of price and performance for your protein separation needs. Novex™ Value Tris-Glycine polyacrylamide gels provide good separation and resolution of your proteins in an affordable gel that is based on traditional Laemmli protein electrophoresis. Each box contains 20 gels.
Features of the Novex Value gels include:
  • Great balance of value and performance
  • Fast run conditions - separate your proteins using constant voltage in less than 60 minutes
  • Improved shelf life - store gels for at least 12 months at 4°C
  • Usable for native and denaturing protein assays
Description & Specifications
Specifications
8cm
8cm
Tris-Glycine Gel
1.0mm