Please login to your online account to display your discounted pricing

WZ 811, Tocris™

Potent CXCR4 antagonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material WZ 811
Assay Percent Range >99%
Molecular Formula C18H18N4
Formula Weight 290.36
Synonym N1, N4-Di-2-pyridinyl-1, 4-benzenedimethanamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

395110

 
r & d systems
RDS395110
10mg Each for $201.36

395150

 
r & d systems
RDS395150
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 290.36
Quantity 10mg
Chemical Name or Material WZ 811
CAS 55778-02-4
Assay Percent Range >99%
Recommended Storage Store at -20°C
Synonym N1, N4-Di-2-pyridinyl-1, 4-benzenedimethanamine
Molecular Formula C18H18N4
Safety and Handling

Store at -20°C