Please login to your online account to display your discounted pricing

WY 45233 succinate, Tocris™

Dual serotonin/noradrenalin re-uptake inhibitor

$246.52 - $1,023.37

Chemical Identifiers

Chemical Name or Material WY 45233 succinate
Assay Percent Range >99%
Molecular Formula C16H25NO2·C4H6O4
Formula Weight 381.46
Synonym 4-[2-(Dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol succinate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

419450

 
r & d systems
RDS419450
50mg Each for $1,023.37

419410

 
r & d systems
RDS419410
10mg Each for $246.52
Description & Specifications

Specifications

Formula Weight 381.46
Quantity 50mg
Synonym 4-[2-(Dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol succinate
Recommended Storage Store at +4°C
Chemical Name or Material WY 45233 succinate
Molecular Formula C16H25NO2·C4H6O4
CAS 448904-47-0
Assay Percent Range >99%
Safety and Handling

Store at +4°C