Please login to your online account to display your discounted pricing

WS 3, Tocris

TRPM8 agonist; cooling agent

$113.57 - $478.51

Chemical Identifiers

Chemical Name or Material WS 3
Assay Percent Range >99%
Molecular Formula C13H25NO
Formula Weight 211.34
Synonym N-Ethyl-5-methyl-2-(1-methylethyl)cyclohexanecarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

292750

 
r & d systems
RDS292750
50mg Each for $478.51

292710

 
r & d systems
RDS292710
10mg Each for $113.57
Description & Specifications

Specifications

Chemical Name or Material WS 3
Quantity 50mg
CAS 39711-79-0
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C13H25NO
Formula Weight 211.34
Synonym N-Ethyl-5-methyl-2-(1-methylethyl)cyclohexanecarboxamide
Safety and Handling

Store at Room Temperature